Assay Details
| Gene Symbol: Pea15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244851.1 | NP_001231780.1 | 3 - 4 | 445 | 98 | |
| GenBank mRNA | AF080001.1 | - | 3 - 4 | 445 | 98 | |
| JI882411.1 | - | 3 - 4 | 467 | 98 | ||
| JP052160.1 | - | 3 - 4 | 467 | 98 | ||
More Information
Set Membership: |
|

Pea15
Genomic Map