Assay Details
| Gene Symbol: B2m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246674.2 | NP_001233603.1 | 3 - 4 | 388 | 119 | |
| GenBank mRNA | JI866227.1 | - | 3 - 4 | 399 | 119 | |
| JP060135.1 | - | 3 - 4 | 399 | 119 | ||
| X57112.1 | - | 3 - 4 | 389 | 119 | ||
More Information
Set Membership: |
|

B2m
Genomic Map