Assay Details
| Gene Symbol: B4galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246695.1 | NP_001233624.1 | 1 - 2 | 328 | 71 | |
| XM_007649755.1 | XP_007647945.1 | 4 - 5 | 845 | 71 | ||
| XM_007649756.1 | XP_007647946.1 | 3 - 4 | 636 | 71 | ||
| XM_007649757.1 | XP_007647947.1 | 3 - 4 | 567 | 71 | ||
| XM_007649758.1 | XP_007647948.1 | 3 - 4 | 750 | 71 | ||
| XM_007649759.1 | XP_007647949.1 | 4 - 5 | 686 | 71 | ||
| GenBank mRNA | AY117538.1 | - | 1 - 2 | 328 | 71 | |
| JI890368.1 | - | 1 - 2 | 399 | 71 | ||
| JP037518.1 | - | 1 - 2 | 399 | 71 | ||
More Information
Set Membership: |
|

B4galt4
Genomic Map