Assay Details
| Gene Symbol: Hspa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246729.1 | NP_001233658.1 | 1 | 252 | 81 | |
| GenBank mRNA | JI867321.1 | - | 1 | 282 | 81 | |
| JP049411.1 | - | 1 | 283 | 81 | ||
| M34561.1 | - | 1 | 252 | 81 | ||
More Information
Set Membership: |
|

Hspa8
Genomic Map