Assay Details
Gene Symbol: Ndufa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246761.1 | NP_001233690.1 | 235 | 74 | ||
GenBank mRNA | AF100706.1 | - | 235 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
