Assay Details
| Gene Symbol: Sepp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256878.1 | NP_001243807.1 | 3 - 4 | 604 | 82 | |
| GenBank mRNA | JI884822.1 | - | 3 - 4 | 604 | 82 | |
| JP052462.1 | - | 3 - 4 | 604 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sepp1| Gene Symbol: Sepp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256878.1 | NP_001243807.1 | 3 - 4 | 604 | 82 | |
| GenBank mRNA | JI884822.1 | - | 3 - 4 | 604 | 82 | |
| JP052462.1 | - | 3 - 4 | 604 | 82 | ||