Assay Details
| Gene Symbol: Gjc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246786.1 | NP_001233715.1 | 1 | 1116 | 85 | |
| XM_007645032.1 | XP_007643222.1 | 6 | 1702 | 85 | ||
| XM_007645033.1 | XP_007643223.1 | 4 | 1488 | 85 | ||
| XM_007645034.1 | XP_007643224.1 | 3 | 1315 | 85 | ||
| GenBank mRNA | AY514680.1 | - | 1 | 1116 | 85 | |
More Information
Set Membership: |
|

Gjc1
Genomic Map