Assay Details
Gene Symbol: Bax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244020.1 | NP_001230949.1 | 3 - 4 | 414 | 67 | |
GenBank mRNA | EF104643.1 | - | 3 - 4 | 414 | 67 | |
JI891563.1 | - | 3 - 4 | 432 | 67 | ||
JP053440.1 | - | 3 - 4 | 432 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
