Assay Details
| Gene Symbol: Casp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244046.1 | NP_001230975.1 | 1 - 2 | 54 | 87 | |
| XM_007641220.1 | XP_007639410.1 | 2 - 3 | 125 | 87 | ||
| XM_007641224.1 | XP_007639414.1 | 2 - 3 | 163 | 87 | ||
| GenBank mRNA | FJ940732.1 | - | 1 - 2 | 54 | 87 | |
| JI896429.1 | - | 2 - 3 | 145 | 87 | ||
| JP037528.1 | - | 2 - 3 | 152 | 87 | ||
More Information
Set Membership: |
|

Casp3
Genomic Map