Assay Details
Gene Symbol: Fis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246714.1 | NP_001233643.1 | 3 - 4 | 251 | 59 | |
GenBank mRNA | AB271736.1 | - | 3 - 4 | 251 | 59 | |
JI875398.1 | - | 3 - 4 | 337 | 59 | ||
JP037636.1 | - | 3 - 4 | 337 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
