Assay Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246789.1 | NP_001233718.1 | 2 - 3 | 377 | 96 | |
| GenBank mRNA | AY649405.1 | - | 2 - 3 | 377 | 96 | |
| JI883463.1 | - | 2 - 3 | 399 | 96 | ||
| JP054321.1 | - | 2 - 3 | 398 | 96 | ||
More Information
Set Membership: |
|

Ndufb11
Genomic Map