Assay Details
| Gene Symbol: Bak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246795.1 | NP_001233724.1 | 3 - 4 | 342 | 60 | |
| XM_007651403.1 | XP_007649593.1 | 4 - 5 | 1178 | 60 | ||
| XM_007651404.1 | XP_007649594.1 | 4 - 5 | 1161 | 60 | ||
| GenBank mRNA | EF104644.1 | - | 3 - 4 | 342 | 60 | |
| JI867360.1 | - | 4 - 5 | 620 | 60 | ||
| JP056774.1 | - | 4 - 5 | 619 | 60 | ||
More Information
Set Membership: |
|

Bak1
Genomic Map