Assay Details
| Gene Symbol: Actb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244575.1 | NP_001231504.1 | 3 - 4 | 807 | 82 | |
| XM_007648665.1 | XP_007646855.1 | 4 - 5 | 906 | 82 | ||
| GenBank mRNA | JI878458.1 | - | 4 - 5 | 901 | 82 | |
| JP040858.1 | - | 2 - 3 | 497 | 82 | ||
More Information
Set Membership: |
|

Actb
Genomic Map