Assay Details
| Gene Symbol: Eef1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244402.1 | NP_001231331.1 | 1 - 2 | 207 | 110 | |
| GenBank mRNA | JI871043.1 | - | 1 - 2 | 210 | 110 | |
| JP062036.1 | - | 1 - 2 | 234 | 110 | ||
More Information
Set Membership: |
|

Eef1a1
Genomic Map