Assay Details
| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244012.1 | NP_001230941.1 | 8 - 9 | 2027 | 128 | |
| GenBank mRNA | AF063024.1 | - | 8 - 9 | 2027 | 128 | |
| JP048748.1 | - | 2 - 3 | 301 | 128 | ||
More Information
Set Membership: |
|

Slc20a1
Genomic Map