Assay Details
| Gene Symbol: Tubb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244097.1 | NP_001231026.1 | 4 | 420 | 89 | |
| XM_007646352.1 | XP_007644542.1 | 4 | 568 | 89 | ||
| GenBank mRNA | JI870787.1 | - | 4 | 523 | 89 | |
| JP038655.1 | - | 4 | 537 | 89 | ||
| X60784.1 | - | 4 | 420 | 89 | ||
More Information
Set Membership: |
|

Tubb
Genomic Map