Assay Details
| Gene Symbol: Tubb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495070.2 | XP_003495118.1 | 3 - 4 | 283 | 108 | |
| XM_007650867.1 | XP_007649057.1 | 3 - 4 | 785 | 108 | ||
More Information
Set Membership: |
|

Tubb3
Genomic Map