Assay Details
Gene Symbol: Pde6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496885.2 | XP_003496933.1 | 2 - 3 | 398 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
