Assay Details
| Gene Symbol: Foxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501385.2 | XP_003501433.1 | 3 - 4 | 765 | 58 | |
| XM_003501386.2 | XP_003501434.1 | 3 - 4 | 1339 | 58 | ||
| XM_007642048.1 | XP_007640238.1 | 3 - 4 | 780 | 58 | ||
| XM_007642050.1 | XP_007640240.1 | 3 - 4 | 820 | 58 | ||
| XM_007642051.1 | XP_007640241.1 | 3 - 4 | 780 | 58 | ||
| XM_007642052.1 | XP_007640242.1 | 3 - 4 | 780 | 58 | ||
More Information
Set Membership: |
|

Foxn3
Genomic Map