Assay Details
Gene Symbol: Krt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511332.2 | XP_003511380.1 | 7 - 8 | 1351 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511332.2 | XP_003511380.1 | 7 - 8 | 1351 | 76 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |