Assay Details
| Gene Symbol: Cela2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652100.1 | XP_007650290.1 | 2 - 3 | 139 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cela2a| Gene Symbol: Cela2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652100.1 | XP_007650290.1 | 2 - 3 | 139 | 73 | |
Set Membership: |
|