Assay Details
| Gene Symbol: Pak6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653056.1 | XP_007651246.1 | 4 - 5 | 1489 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pak6| Gene Symbol: Pak6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653056.1 | XP_007651246.1 | 4 - 5 | 1489 | 66 | |
Set Membership: |
|