Assay Details
Gene Symbol: Wasf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651967.1 | XP_007650157.1 | 3 - 4 | 327 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
