Assay Details
Gene Symbol: LOC100765482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654040.1 | XP_007652230.1 | 5 - 6 | 627 | 70 | |
GenBank mRNA | JI871498.1 | - | 4 - 5 | 448 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
