Assay Details
Gene Symbol: LOC100765482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654040.1 | XP_007652230.1 | 7 - 8 | 878 | 84 | |
GenBank mRNA | JI871498.1 | - | 6 - 7 | 699 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
