Assay Details
| Gene Symbol: Kdelr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652679.1 | XP_007650869.1 | 784 | 77 | ||
| GenBank mRNA | JI878718.1 | - | 784 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdelr1| Gene Symbol: Kdelr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652679.1 | XP_007650869.1 | 784 | 77 | ||
| GenBank mRNA | JI878718.1 | - | 784 | 77 | ||
Set Membership: |
|