Assay Details
Gene Symbol: Ufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506861.2 | XP_003506909.1 | 2 - 3 | 450 | 64 | |
GenBank mRNA | JI882634.1 | - | 2 - 3 | 418 | 64 | |
JP061253.1 | - | 2 - 3 | 418 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
