Assay Details
| Gene Symbol: Tuba4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507592.2 | XP_003507640.1 | 3 - 4 | 444 | 91 | |
| XM_007648186.1 | XP_007646376.1 | 3 - 4 | 507 | 91 | ||
| XM_007648187.1 | XP_007646377.1 | 3 - 4 | 823 | 91 | ||
| XM_007648188.1 | XP_007646378.1 | 3 - 4 | 469 | 91 | ||
More Information
Set Membership: |
|

Tuba4a
Genomic Map