Assay Details
| Gene Symbol: Pycrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510442.2 | XP_003510490.2 | 2 - 3 | 363 | 65 | |
| GenBank mRNA | JP057295.1 | - | 2 - 3 | 177 | 65 | |
More Information
Set Membership: |
|

Pycrl
Genomic Map