Assay Details
| Gene Symbol: Pycrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510442.2 | XP_003510490.2 | 3 - 4 | 543 | 94 | |
| GenBank mRNA | JP057295.1 | - | 3 - 4 | 357 | 94 | |
More Information
Set Membership: |
|

Pycrl
Genomic Map