Assay Details
Gene Symbol: Bcl2l12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654274.1 | XP_007652464.1 | 3 - 4 | 334 | 59 | |
GenBank mRNA | JI892672.1 | - | 3 - 4 | 373 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
