Assay Details
Gene Symbol: LOC100773414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501803.1 | XP_003501851.1 | 3 - 4 | 458 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
