Assay Details
Gene Symbol: Etv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502894.2 | XP_003502942.1 | 3 - 4 | 701 | 84 | |
XM_007643495.1 | XP_007641685.1 | 3 - 4 | 354 | 84 | ||
XM_007643496.1 | XP_007641686.1 | 3 - 4 | 354 | 84 | ||
GenBank mRNA | JI867058.1 | - | 1 - 2 | 114 | 84 | |
JP039416.1 | - | 1 - 2 | 114 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
