Assay Details
| Gene Symbol: Prss22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650149.1 | XP_007648339.1 | 2 - 3 | 242 | 73 | |
| XM_007650150.1 | XP_007648340.1 | 2 - 3 | 1103 | 73 | ||
More Information
Set Membership: |
|

Prss22
Genomic Map