Assay Details
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652539.1 | XP_007650729.1 | 6 - 7 | 1049 | 62 | |
XM_007652540.1 | XP_007650730.1 | 6 - 7 | 1038 | 62 | ||
XM_007652541.1 | XP_007650731.1 | 6 - 7 | 1043 | 62 | ||
XM_007652542.1 | XP_007650732.1 | 6 - 7 | 1049 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
