Assay Details
| Gene Symbol: Tmed3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495623.2 | XP_003495671.1 | 1 - 2 | 306 | 72 | |
| GenBank mRNA | JI877111.1 | - | 1 - 2 | 316 | 72 | |
| JP045898.1 | - | 1 - 2 | 316 | 72 | ||
More Information
Set Membership: |
|

Tmed3
Genomic Map