Assay Details
Gene Symbol: Ndufaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496208.2 | XP_003496256.1 | 2 - 3 | 264 | 73 | |
GenBank mRNA | JI871748.1 | - | 1 - 2 | 87 | 73 | |
JP066059.1 | - | 1 - 2 | 85 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
