Assay Details
Gene Symbol: Dnajc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497013.2 | XP_003497061.1 | 6 - 7 | 853 | 64 | |
GenBank mRNA | JI879123.1 | - | 6 - 7 | 767 | 64 | |
JP052568.1 | - | 6 - 7 | 767 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
