Assay Details
| Gene Symbol: Atp5g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498323.2 | XP_003498371.1 | 2 - 3 | 253 | 81 | |
| GenBank mRNA | JI896039.1 | - | 2 - 3 | 253 | 81 | |
| JP060363.1 | - | 2 - 3 | 253 | 81 | ||
More Information
Set Membership: |
|

Atp5g3
Genomic Map