Assay Details
Gene Symbol: Timm17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505098.1 | XP_003505146.1 | 3 - 4 | 253 | 68 | |
GenBank mRNA | JI875859.1 | - | 3 - 4 | 259 | 68 | |
JP060529.1 | - | 3 - 4 | 259 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
