Assay Details
| Gene Symbol: Surf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511727.2 | XP_003511775.1 | 6 - 7 | 612 | 67 | |
| XM_007652258.1 | XP_007650448.1 | 5 - 6 | 538 | 67 | ||
| GenBank mRNA | JI863065.1 | - | 2 - 3 | 150 | 67 | |
More Information
Set Membership: |
|

Surf1
Genomic Map