Assay Details
| Gene Symbol: Atp6v0c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650145.1 | XP_007648335.1 | 2 - 3 | 462 | 84 | |
| GenBank mRNA | JI889332.1 | - | 2 - 3 | 421 | 84 | |
| JP054425.1 | - | 2 - 3 | 421 | 84 | ||
More Information
Set Membership: |
|

Atp6v0c
Genomic Map