Assay Details
Gene Symbol: Cdkn2aipnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007628676.1 | XP_007626866.1 | 1 - 2 | 401 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
