Assay Details
Gene Symbol: LOC100756827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501827.2 | XP_003501875.2 | 5 - 6 | 600 | 80 | |
XM_007642459.1 | XP_007640649.1 | 6 - 7 | 677 | 80 | ||
XM_007642460.1 | XP_007640650.1 | 5 - 6 | 600 | 80 | ||
XM_007642461.1 | XP_007640651.1 | 5 - 6 | 600 | 80 | ||
XM_007642462.1 | XP_007640652.1 | 5 - 6 | 600 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
