Assay Details
Gene Symbol: Slc51a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495400.2 | XP_003495448.1 | 4 - 5 | 702 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
