Assay Details
| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496275.2 | XP_003496323.1 | 2 - 3 | 913 | 82 | |
| XM_007648676.1 | XP_007646866.1 | 2 - 3 | 916 | 82 | ||
More Information
Set Membership: |
|

Ankrd61
Genomic Map