Assay Details
| Gene Symbol: Fhdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505738.2 | XP_003505786.1 | 10 - 11 | 1455 | 69 | |
| XM_007646214.1 | XP_007644404.1 | 10 - 11 | 1405 | 69 | ||
| XM_007646215.1 | XP_007644405.1 | 10 - 11 | 1534 | 69 | ||
More Information
Set Membership: |
|

Fhdc1
Genomic Map