Assay Details
Gene Symbol: Minos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506890.2 | XP_003506938.1 | 1 - 2 | 172 | 102 | |
GenBank mRNA | JI883523.1 | - | 1 - 2 | 154 | 102 | |
JP063870.1 | - | 1 - 2 | 154 | 102 |

More Information
Set Membership: |
![]() ![]() |
