Assay Details
Gene Symbol: Nr0b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507919.1 | XP_003507967.1 | 1 - 2 | 1164 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
