Assay Details
Gene Symbol: Pou1f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512830.1 | XP_003512878.1 | 5 - 6 | 668 | 78 | |
XM_003512831.2 | XP_003512879.1 | 5 - 6 | 782 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
